• USA Home
  • Search Results

  • nndimethylethylenediamine881510800911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Dimethylethylenediamine

3 Products
Synonym:   2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane
Image of N,N-<WBR>Dimethylethylenediamine
CAS Number:
Linear Formula:
 NH2C2H4N(CH3)2
Molecular Weight:
 88.15
 EC Index Number
 612-075-00-2
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More