• USA Home
  • Search Results

  • nndimethylpphenylenediaminedihydrochloride2091253646911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Dimethyl-p-phenylenediamine dihydrochloride

2 Products
Synonym:   4-Amino-N,N-dimethylaniline dihydrochloride, DMPD · 2HCl, DMPPDA · 2HCl
Image of <I>N</I>,<I>N</I>-Dimethyl-<I>p</I>-phenylenediamine dihydrochloride
CAS Number:
Linear Formula:
 (CH3)2NC6H4NH2 · 2HCl
Molecular Weight:
 209.12
 Beilstein/REAXYS Number:
 3913463
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More