• USA Home
  • Search Results

  • nndimethylpphenylenediaminesulfatesalt2342753647011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Dimethyl-p-phenylenediamine sulfate salt

1 Products
Synonym:   4-(Dimethylamino)aniline sulfate salt, 4-Amino-N,N-dimethylaniline sulfate salt, DMPPDA
Image of <I>N,N</I>-Dimethyl-<I>p</I>-phenylenediamine sulfate salt
CAS Number:
Linear Formula:
 (CH3)2NC6H4NH2·H2SO4
Molecular Weight:
 234.27
 Beilstein/REAXYS Number:
 4612278
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More