• USA Home
  • Search Results

  • nndimethyltetradecylamine2414611275411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N,N-Dimethyltetradecylamine

1 Products
Synonym:   1-(Dimethylamino)tetradecane, N,N-Dimethylmyristylamine
Image of <I>N</I>,<I>N</I>-<WBR>Dimethyltetradecylamine
CAS Number:
Linear Formula:
 CH3(CH2)13N(CH3)2
Molecular Weight:
 241.46
 Beilstein/REAXYS Number:
 1748246
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More