• USA Home
  • Search Results

  • nonacosane4087963003511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nonacosane

2 Products
Image of Nonacosane
CAS Number:
Linear Formula:
 CH3(CH2)27CH3
Molecular Weight:
 408.79
 Beilstein/REAXYS Number:
 1724922
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More