• USA Home
  • Search Results

  • nonane1282611184211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nonane

4 Products
Synonym:   n-Nonane
Image of Nonane
CAS Number:
Linear Formula:
 CH3(CH2)7CH3
Molecular Weight:
 128.26
 Beilstein/REAXYS Number:
 1696917
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Assay
  Special Grade
Show More