• USA Home
  • Search Results

  • nonanoicacid1582411205011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nonanoic acid

6 Products
Synonym:   Acid C9, Pelargonic acid
Image of Nonanoic acid
CAS Number:
Linear Formula:
 CH3(CH2)7COOH
Molecular Weight:
 158.24
 Beilstein/REAXYS Number:
 1752351
 Council of Europe no.:
 29
 EC Number:
 FEMA Number:
 2784
 Flavis number:
 8.029
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Functional group
  Allergen
  Organoleptic property
  Physical Form
  Application(s)
  Ice Shipping
  Biological Source
  Special Grade
  Agency/Method
  Assay
Show More