• USA Home
  • Search Results

  • nonylbdglucopyranoside306406998473211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nonyl β-D-glucopyranoside

1 Products
Image of Nonyl β-<SC>D</SC>-glucopyranoside
CAS Number:
Empirical Formula (Hill Notation):
 C15H30O6
Molecular Weight:
 306.40
 Beilstein/REAXYS Number:
 12168
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More