• USA Home
  • Search Results

  • nootkatone21833467450411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nootkatone

1 Products
Synonym:   (+)-Nootkatone
Image of Nootkatone
CAS Number:
Empirical Formula (Hill Notation):
 C15H22O
Molecular Weight:
 218.33
 Beilstein/REAXYS Number:
 4676969
 EC Number:
 FEMA Number:
 3166
 Flavis number:
 07.089
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
Show More

Product #
Description
W316620 Nootkatone, ≥98%, FG,