• USA Home
  • Search Results

  • nordihydroguaiareticacid3023650038911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nordihydroguaiaretic acid

3 Products
Synonym:   1,4-Bis(3,4-dihydroxyphenyl)-2,3-dimethylbutane, 4,4′-(2,3-Dimethyltetramethylene)dipyrocatechol, Masoprocol, NDGA
Image of Nordihydroguaiaretic acid
CAS Number:
Linear Formula:
 [-CH(CH3)CH2C6H3-1,2-(OH)2]2
Molecular Weight:
 302.36
 Beilstein/REAXYS Number:
 2056826
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More