• USA Home
  • Search Results

  • norgestimate369503518928711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Norgestimate

5 Products
Synonym:   (17α)-17-(Acetyloxy)-13-ethyl-18,19-dinorpregn-4-en-20-yn-3-one 3-oxime, Dexnorgestrel acetime
Image of Norgestimate
CAS Number:
Empirical Formula (Hill Notation):
 C23H31NO3
Molecular Weight:
 369.50
 Beilstein/REAXYS Number
 6440219
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Titertest
  Assay
  Physical Form
Show More