• USA Home
  • Search Results

  • nornicotine14820574686111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Nornicotine

1 Products
Synonym:   2-(3-Pyridyl)pyrrolidine, 3-(2-Pyrrolidinyl)pyridine
Image of (±)-Nornicotine
CAS Number:
Empirical Formula (Hill Notation):
 C9H12N2
Molecular Weight:
 148.20
 Beilstein/REAXYS Number:
 81968
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
 eCl@ss:
 39151701
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More