• USA Home
  • Search Results

  • novobiocinsodiumsalt63461147653511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Novobiocin sodium salt

3 Products
Image of Novobiocin sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C31H35N2NaO11
Molecular Weight:
 634.61
 Beilstein/REAXYS Number:
 3892910
 EC Number:
 NACRES:
 NA.85
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Mode of Action
  Physical Form
  Application(s)
  Solubility
  Activity Spectrum
  Assay
Show More