• USA Home
  • Search Results

  • nphenylanthranilicacid213239140711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Phenylanthranilic acid

2 Products
Synonym:   2-(Phenylamino)benzoic acid, Diphenylamine-2-carboxylic acid, DpC
Image of <I>N</I>-Phenylanthranilic acid
CAS Number:
Linear Formula:
 2-(C6H5NH)C6H4CO2H
Molecular Weight:
 213.23
 Beilstein/REAXYS Number:
 1456607
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Melting Point
Show More