• USA Home
  • Search Results

  • nphenylglycine1511610301511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-Phenylglycine

1 Products
Synonym:   (Phenylamino)acetic acid, Anilinoacetic acid
Image of <I>N</I>-Phenylglycine
CAS Number:
Linear Formula:
 C6H5NHCH2COOH
Molecular Weight:
 151.16
 Beilstein/REAXYS Number:
 509838
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
330469 N-Phenylglycine, 97%,