• USA Home
  • Search Results

  • nptosyllphenylalaninechloromethylketone3518540271111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-p-Tosyl-L-phenylalanine chloromethyl ketone

1 Products
Synonym:   (S)-1-Chloro-3-tosylamido-4-phenyl-2-butanone, (S)-1-Chloro-4-phenyl-3-tosylamido-2-butanone, TPCK, Tosyl-L-phenylalanyl-chloromethane
Image of N-<I>p</I>-Tosyl-<SC>L</SC>-phenylalanine chloromethyl ketone
CAS Number:
Empirical Formula (Hill Notation):
 C17H18ClNO3S
Molecular Weight:
 351.85
 Beilstein/REAXYS Number:
 2895215
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More