• USA Home
  • Search Results

  • ntertbutyldiethanolamine16124216093211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-tert-Butyldiethanolamine

1 Products
Synonym:   N,N-Bis(2-hydroxyethyl)-tert-butylamine, N-tert-Butyl-2,2′-iminodiethanol
Image of <I>N</I>-<I>tert</I>-Butyldiethanolamine
CAS Number:
Linear Formula:
 (CH3)3CN(CH2CH2OH)2
Molecular Weight:
 161.24
 Beilstein/REAXYS Number:
 1739942
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More