• USA Home
  • Search Results

  • ntrishydroxymethylmethylacrylamide175181388005211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

N-[Tris(hydroxymethyl)methyl]acrylamide

1 Products
Synonym:   NAT, TRIS-acrylamide
Image of <I>N</I>-<WBR>[Tris(hydroxymethyl)<WBR>methyl]<WBR>acrylamide
CAS Number:
Linear Formula:
 H2C=CHCONHC(CH2OH)3
Molecular Weight:
 175.18
 Beilstein/REAXYS Number:
 2081162
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More