• USA Home
  • Search Results

  • nuclearfastred35727640977411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nuclear Fast Red

4 Products
Synonym:   4-Amino-9,10-dihydro-1,3-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt
Image of Nuclear Fast Red
CAS Number:
Empirical Formula (Hill Notation):
 C14H8NNaO7S
Molecular Weight:
 357.27
 Colour Index Number:
 60760
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More