• USA Home
  • Search Results

  • nucleasemicrococcalfromstaphylococcusaureus12345901353011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nuclease micrococcal from Staphylococcus aureus

3 Products
Synonym:   Endonuclease micrococcal, MNase, Micrococcal endonuclease, Micrococcal nuclease
Image of Nuclease micrococcal from <I>Staphylococcus aureus</I>
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.1.31.1  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Specific Activity
  Physical Form
  Chemical Composition
Show More