• USA Home
  • Search Results

  • nylon612123452609855511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Nylon 6/12

1 Products
Synonym:   Hexamethylenediamine-1,12-dodecanedioic acid polymer, Poly(hexamethylene dodecanediamide)
Image of Nylon 6/12
CAS Number:
Linear Formula:
 (C12H22O4 · C6H16N2)n
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Transition temperature
Show More