• USA Home
  • Search Results

  • o23mercaptopropionylaminoethylomethylpolyethyleneglycol1234540191661811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

O-[2-(3-Mercaptopropionylamino)ethyl]-O′-methylpolyethylene glycol

1 Products
Synonym:   ‘Mercaptopolyethylene glycol monomethyl ether’, PEG-thiol
Image of <I>O</I>-<WBR>[2-<WBR>(3-<WBR>Mercaptopropionylamino)<WBR>ethyl]<WBR>-<I>O</I>′-methylpolyethylene glycol
CAS Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More