• USA Home
  • Search Results

  • o2oxo12hpyridylnnnntetramethyluroniumtetrafluoroborate2970612570071211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

O-(2-Oxo-1(2H)pyridyl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate

1 Products
Synonym:   O-(1,2-Dihydro-2-oxo-1-pyridyl)-N,N,N′-N′-tetramethyluronium tetrafluoroborate, TPTU
Image of <I>O</I>-(2-Oxo-1(2<I>H</I>)pyridyl)-<I>N</I>,<I>N</I>,<I>N</I>′,<I>N</I>′-tetramethyluronium tetrafluoroborate
CAS Number:
Empirical Formula (Hill Notation):
 C10H16BF4N3O2
Molecular Weight:
 297.06
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More