• USA Home
  • Search Results

  • oacetylsalicyloylchloride19860553851211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

O-Acetylsalicyloyl chloride

1 Products
Image of <I>O</I>-Acetylsalicyloyl chloride
CAS Number:
Linear Formula:
 2-(CH3CO2)C6H4COCl
Molecular Weight:
 198.60
 Beilstein/REAXYS Number:
 880372
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More