• USA Home
  • Search Results

  • oanisaldehyde1361513502411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

o-Anisaldehyde

2 Products
Synonym:   2-Methoxybenzaldehyde, o-Anisaldehyde, Salicylaldehyde methyl ether
Image of <I>o</I>-Anisaldehyde
CAS Number:
Linear Formula:
 CH3OC6H4CHO
Molecular Weight:
 136.15
 Beilstein/REAXYS Number:
 606301
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Organoleptic property
  Biological Source
  Assay
  Special Grade
Show More