• USA Home
  • Search Results

  • oanisidine123159004011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

o-Anisidine

2 Products
Synonym:   2-Aminoanisole, 2-Methoxyaniline
Image of <I>o</I>-Anisidine
CAS Number:
Linear Formula:
 CH3OC6H4NH2
Molecular Weight:
 123.15
 Beilstein/REAXYS Number:
 386210
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More