• USA Home
  • Search Results

  • obenzotriazol1ylnnnnbistetramethyleneuroniumhexafluorophosphate4313210537924611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

O-(Benzotriazol-1-yl)-N,N,N′,N′-bis(tetramethylene)uronium hexafluorophosphate

1 Products
Synonym:   (Benzotriazol-1-yloxy)dipyrrolidinocarbenium hexafluorophosphate, HBPyU
Image of <I>O</I>-<WBR>(Benzotriazol-<WBR>1-<WBR>yl)<WBR>-<I>N,N,N′,N′</I>-bis(tetramethylene)uronium hexafluorophosphate
CAS Number:
Empirical Formula (Hill Notation):
 C15H20F6N5OP
Molecular Weight:
 431.32
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More