• USA Home
  • Search Results

  • obenzylhydroxylaminehydrochloride15961268743611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

O-Benzylhydroxylamine hydrochloride

1 Products
Synonym:   Benzyloxyamine hydrochloride
Image of <I>O</I>-Benzylhydroxylamine hydrochloride
CAS Number:
Linear Formula:
 C6H5CH2ONH2 · HCl
Molecular Weight:
 159.61
 Beilstein/REAXYS Number:
 3687991
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More