• USA Home
  • Search Results

  • obenzylltyrosine271311665264511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

O-Benzyl-L-tyrosine

1 Products
Synonym:   4-Benzyloxy-L-phenylalanine
Image of <I>O</I>-Benzyl-<SC>L</SC>-tyrosine
CAS Number:
Linear Formula:
 C6H5CH2OC6H4CH2CH(NH2)CO2H
Molecular Weight:
 271.31
 Beilstein/REAXYS Number:
 2659642
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More