• USA Home
  • Search Results

  • ochratoxinabsaconjugatefromaspergillusochraceus1234530347911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ochratoxin A−BSA conjugate from Aspergillus ochraceus

1 Products
Image of Ochratoxin A−BSA conjugate from <I>Aspergillus ochraceus</I>
CAS Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More