• USA Home
  • Search Results

  • ocimene136231387791311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ocimene

1 Products
Synonym:   3,7-Dimethyl-1,3,6-octatrien, 3,7-Dimethyl-1,3,6-octatriene
Image of Ocimene
CAS Number:
Empirical Formula (Hill Notation):
 C10H16
Molecular Weight:
 136.23
 Beilstein/REAXYS Number:
 1736172
 EC Number:
 FEMA Number:
 3539
 Flavis number:
 1.018
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More