• USA Home
  • Search Results

  • ocresol108149548711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

o-Cresol

4 Products
Synonym:   2-Methylphenol
Image of <I>o</I>-Cresol
CAS Number:
Linear Formula:
 CH3C6H4OH
Molecular Weight:
 108.14
 Beilstein/REAXYS Number:
 506917
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Vapor pressure
  Application(s)
  Potency
  Density
  Special Grade
  pH Value
  Assay
Show More