• USA Home
  • Search Results

  • octadecyl335ditertbutyl4hydroxyphenylpropionate53086208279311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate

3 Products
Image of Octadecyl 3-(3,5-di-<I>tert</I>-<WBR>butyl-<WBR>4-<WBR>hydroxyphenyl)<WBR>propionate
CAS Number:
Linear Formula:
 [(CH3)3C]2C6H2(OH)CH2CH2CO2(CH2)17CH3
Molecular Weight:
 530.86
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
Show More