• USA Home
  • Search Results

  • octamethylcyclotetrasiloxane2966255667211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Octamethylcyclotetrasiloxane

3 Products
Image of Octamethylcyclotetrasiloxane
CAS Number:
Linear Formula:
 [-Si(CH3)2O-]4
Molecular Weight:
 296.62
 Beilstein/REAXYS Number:
 1787074
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Potency
Show More