• USA Home
  • Search Results

  • octane1142311165911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Octane

5 Products
Synonym:   n-Octane
Image of Octane
CAS Number:
Linear Formula:
 CH3(CH2)6CH3
Molecular Weight:
 114.23
 Beilstein/REAXYS Number:
 1696875
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Solubility
  Special Grade
Show More