• USA Home
  • Search Results

  • octocrylene36148619730411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Octocrylene

3 Products
Synonym:   2-Ethylhexyl 2-cyano-3,3-diphenylacrylate
Image of Octocrylene
CAS Number:
Linear Formula:
 (C6H5)2C=C(CN)CO2CH2CH(C2H5)(CH2)3CH3
Molecular Weight:
 361.48
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More