• USA Home
  • Search Results

  • octylbdglucopyranoside292372983626811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Octyl β-D-glucopyranoside

2 Products
Synonym:   n-Octyl glucoside, OGP
Image of Octyl β-<SC>D</SC>-glucopyranoside
CAS Number:
Empirical Formula (Hill Notation):
 C14H28O6
Molecular Weight:
 292.37
 Beilstein/REAXYS Number:
 84118
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More