• USA Home
  • Search Results

  • ofloxacin361378241936111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ofloxacin

4 Products
CAS Number:
Empirical Formula (Hill Notation):
 C18H20FN3O4
Molecular Weight:
 361.37
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Mode of Action
  Titertest
  Activity Spectrum
  Anal. Standard Form
  Physical Form
Show More