• USA Home
  • Search Results

  • oilredegn39447447779611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oil Red EGN

1 Products
Synonym:   Solvent Red 26
Image of Oil Red EGN
CAS Number:
Empirical Formula (Hill Notation):
 C25H22N4O
Molecular Weight:
 394.47
 Colour Index Number:
 26120
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
234117 Oil Red EGN,