• USA Home
  • Search Results

  • oilredosolution40849132006511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oil Red O solution

2 Products
Synonym:   Solvent Red 27, Sudan Red 5B
Image of Oil Red O solution
CAS Number:
Empirical Formula (Hill Notation):
 C26H24N4O
Molecular Weight:
 408.49
 EC Number:
 NACRES:
 NA.47
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Absorption
  Physical Form
  Concentration
Show More