• USA Home
  • Search Results

  • oleamide2814830102011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oleamide

3 Products
Synonym:   cis-9,10-Octadecenoamide, cis-9-Octadecenamide, Oleic acid amide
Image of Oleamide
CAS Number:
Empirical Formula (Hill Notation):
 C18H35NO
Molecular Weight:
 281.48
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
Show More