• USA Home
  • Search Results

  • oleicacid2824611280111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oleic acid

11 Products
Synonym:   cis-9-Octadecenoic acid, Elainic acid
Image of Oleic acid
CAS Number:
Linear Formula:
 CH3(CH2)7CH=CH(CH2)7COOH
Molecular Weight:
 282.46
 Beilstein/REAXYS Number:
 1726542
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Quality clarifier
  Product Line
  Manufacturer designation
  Anal. Standard Form
  Shelf Life
  Suitability
  Density
  Biological Source
  Ice Shipping
  Agency/Method
  Functional group
  Application(s)
  Chemical Composition
  Solubility
  Physical Form
  Assay
Show More