• USA Home
  • Search Results

  • oleoylchloride3009111277611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oleoyl chloride

1 Products
Image of Oleoyl chloride
CAS Number:
Linear Formula:
 CH3(CH2)7CH=CH(CH2)7COCl
Molecular Weight:
 300.91
 Beilstein/REAXYS Number:
 1211748
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
367850 Oleoyl chloride, ≥89%,