• USA Home
  • Search Results

  • oleylalcohol2684814328211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oleyl alcohol

5 Products
Synonym:   Z-9-Dodecen-1-ol, cis-9-Octadecen-1-ol
Image of Oleyl alcohol
CAS Number:
Linear Formula:
 CH3(CH2)7CH=CH(CH2)7CH2OH
Molecular Weight:
 268.48
 Beilstein/REAXYS Number:
 1723962
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More