• USA Home
  • Search Results

  • olmesartanmedoxomil5585914468963411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Olmesartan medoxomil

5 Products
Synonym:   4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester, CS-866
Image of Olmesartan medoxomil
CAS Number:
Empirical Formula (Hill Notation):
 C29H30N6O6
Molecular Weight:
 558.59
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Manufacturer designation
Show More