• USA Home
  • Search Results

  • omnipurwaterdepctreatedsterilenucleasefreecas7732185calbiochem1802773218511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

OmniPur Water, DEPC Treated, Sterile, Nuclease-Free - CAS 7732-18-5 - Calbiochem

1 Products
Synonym:   Water
Image of OmniPur<TMSYMBOL></TMSYMBOL> Water, DEPC Treated, Sterile, Nuclease-Free - CAS 7732-18-5 - Calbiochem
CAS Number:
Linear Formula:
 H2O
Molecular Weight:
 18.02
 Beilstein/REAXYS Number:
 2050024
 MDL number:
 NACRES
 NA.26
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More