• USA Home
  • Search Results

  • ondansetronhydrochloride3658510363904911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ondansetron hydrochloride

2 Products
Synonym:   1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one hydrochloride, GR 38032F, Ondansetron hydrochloride dihydrate
Image of Ondansetron hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C18H19N3O · HCl · 2H2O
Molecular Weight:
 365.85
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
Show More