• USA Home
  • Search Results

  • oodiethylchlorothiophosphate18861252404111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

O,O′-Diethyl chlorothiophosphate

1 Products
Synonym:   O,O′-Diethylthiophosphoric chloride, Diethyl phosphorochloridothionate
Image of <I>O,O′</I>-Diethyl chlorothiophosphate
CAS Number:
Linear Formula:
 (CH3CH2O)2P=(S)Cl
Molecular Weight:
 188.61
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More