• USA Home
  • Search Results

  • orangeiisodiumsalt3503263396511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Orange II sodium salt

3 Products
Synonym:   4-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid sodium salt, Acid Orange 7, Acid Orange A, Orange II, Tropaeolin 000 No. 2
Image of Orange II sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C16H11N2NaO4S
Molecular Weight:
 350.32
 Beilstein/REAXYS Number:
 3898201
 Colour Index Number:
 15510
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Chemical Composition
  Titertest
Show More