• USA Home
  • Search Results

  • oroxylina2842648011511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Oroxylin A

1 Products
Synonym:   5,7-Dihydroxy-6-methoxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-6-methoxyflavone;, 6-Methoxybaicalein;
Image of Oroxylin A
CAS Number:
Empirical Formula (Hill Notation):
 C16H12O5
Molecular Weight:
 284.26
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More